Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Forensic Enzymology Kit
SDPSCI_ED

Catalog No. s99518 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S99518 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S99518 Supplier Edvotek™ Supplier No. 193
Only null left
Add to Cart
Add to Cart

Includes

Enough supplies for 10 student groups

Can enzymes be used as evidence?

In a head-on accident, both drivers claimed that the other driver fell asleep at the wheel and caused a fatal accident which was later ruled a homicide. The attending physician at the local hospital did a thorough examination of the two drivers, took blood and urine samples and measured the drivers' temperatures using a disposable plastic mouthpiece and a tongue depressor, which he saved.

Fortunately, the physician had read that the level of saliva amylase increases in humans who are sleep deprived. In this exercise, students will:

  • Analyze the level of saliva amylase found on the tongue depressors of the two drivers
  • Use the results to determine who fell asleep at the wheel, causing the accident

This experiment does not contain components that have been prepared from human sources.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Analysis saliva amylase to determine which fictional driver fall asleep
Includes Instructions, simulated control and driver saliva samples, starch, hcl, iodine, and detection solutions, transfer pipets, microtiter plates, microtest tubes, redeemable coupon for refrigerated items
Requires Visible wavelength spectrophotometer, test tube racks, lab permanent markers, test tubes, beakers, distilled water, linear graph paper
Content And Storage Refrigerated
Class Size 10 Lab Groups
Product Type Forensics Blood Typing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.