Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ How Clean Is the Water We Drink and the Air We Breathe?
SDPSCI_ED

Catalog No. S94599 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94599 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94599 Supplier Edvotek™ Supplier No. S30
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Two bottles of ReadyPour™ agar; Three sleeves Petri plates; Two 10mL pipets; Pack of 1mL pipets; 20mL sterile water sample

Requires

Water samples; Test tubes; Pipet pump or bulb; Hotplate or water bath; Aluminum foil or plastic wrap; 10% bleach solution

Learn about microbes present in our everyday environment.

  • Bacterial concentrations, referred to as bacterial counts, vary depending on the source of water or air
  • Test and compare bacterial counts from various air and water sources
  • All components are stable at room temperature
  • For 10 lab groups

Order Info

Class size: 30 students working in pairs.

Specifications

Grade 6 to 12
Includes Materials for 10 lab groups
Class Size 10 Lab Groups
Product Type Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.