Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ How Does a Doctor Test for AIDS?
SDPSCI_ED

Catalog No. S94603 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94603 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94603 Supplier Edvotek™ Supplier No. S70
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Simulated HIV antigens
  • Positive control
  • Negative control
  • Donor 1 serum
  • Donor 2 serum
  • Secondary antibody (2°Ab)
  • Substrate
  • HIV antibody detection strips
  • Small transfer pipets
  • Large transfer pipets
  • Microtest tubes
  • Complete instructions, background information and study questions

Requires

  • Distilled water
  • Beakers, 50 or 100mL
  • Pencils
  • Gloves
  • Timer or clock

Students perform simulated ELISA testing for AIDS.

  • Immunology-based diagnostic tests are used to detect various diseases
  • Enzyme Linked Immunosorbent Assay (ELISA) tests for antibodies in the blood which indicate infection
  • Students perform a simulated ELISA test to identify 'infected' samples and compare them to control samples
  • Complete in 45 minutes

No human blood or components are used in this experiment.

Order Info

For 10 groups.

Specifications

Grade 6 to 12
For Use With (Application) Students perform simulated elisa testing for aids
Includes Instructions, antigens, positive and negative controls, sera, secondary antibody, substrate, detection strips, transfer pipets and test tubes
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type How Does A Doctor Test for Aids?
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.