Learn More
Edvotek™ In Search of Cancer Gene
SDPSCI_ED
Catalog No. S11314ND Shop All EDVOTEK Products
Change view
Click to view available options
For Use With (Equipment):
Electrophoresis Apparatus
Catalog No. For Use With (Equipment)
S11314ND Electrophoresis Apparatus
1 options
Catalog No. S11314ND Supplier Edvotek™ Supplier No. 314
Add to Cart
Add to Cart
Includes
instructions, Ready-to-load™ Predigested DNA samples, UltraSpec-Agarose™ powder, electrophoresis buffer, InstaStain™ Ethidium Bromide, 5 autoradiograms
Students will get to learn the effect of mutations in p53 tumor suppressor gene and its role in familial cancers.
Suppressor genes such as p53 are essential for cell functions. Mutations in the p53 gene can be correlated to predisposition for certain cancers. Mutations in genes can either be inherited or accumulated due to environmental insults. This experiment deals with a family pedigree determination of several generations relating to cancer formation due to p53 gene mutation. This experiment does not contain human DNA.
Specifications
For Use With (Equipment) Electrophoresis Apparatus
For Use With (Application) For use students will get to learn the effect of mutations in p53 tumor suppressor gene and its role in familial cancers
Includes Instructions, ready to load, predigested DNA samples, ultraSpec Agarose powder, practice Gel loading solution, electrophoresis Buffer, InstaStain™ Ethidium Bromide, pipet, 5 autoradiograms
Product Type Electrophoresis Kit
Quantity 10 Groups