Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ In Search of My Father
SDPSCI_ED

Catalog No. S05713 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05713 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05713 Supplier Edvotek™ Supplier No. S49
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Ready-to-Load™ dye samples
  • Agarose powder
  • Practice gel-loading solution
  • Buffer
  • Microtipped transfer pipets

Requires

  • Electrophoresis apparatus
  • Power supply
  • Microwave or hotplate

Solve the mystery of two boys separated from their parents a decade ago.

  • Students identify the biological mother by mitochondrial DNA
  • Identify the biological father using chromosomal DNA
  • Complete in 45 minutes

Order Info

For 10 lab groups.

Specifications

Grade 6 to 12
For Use With (Application) For determining paternity/maternity
Includes Instructions, ready-to-load™ dye samples, agarose powder, practice gel loading solution, buffer, microtipped transfer pipets
Requires Electrophoresis apparatus, power supply, microwave or hot plate
Time Range 45 min.
Content And Storage Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.
Class Size 10 Lab Groups
Product Type Paternity/Maternity Testing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.