Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ In Search of the Sickle Cell Gene by Southern Blot
SDPSCI_ED

Catalog No. S11315ND Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
5 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11315ND 5 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11315ND Supplier Edvotek™ Supplier No. 315
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, Ready-to-Load™ DNA samples, agarose, electrophoresis buffer, nylon membranes, filter paper, blot stain

Requires

  • Horizontal gel electrophoresis apparatus
  • Hotplate
  • Micropipets
  • Balance
  • Water bath
  • Incubation oven
  • 250mL Flasks
  • All necessary safety equipment
  • Concentrated HCl
  • NaOH
  • Forceps Distilled or deionized water

Use Southern blots to detect mutations while gaining insight about the effects of sickle cell anemia.

Southern blotting is an important technique used widely in clinical genetics and research. By transferring DNA from an agarose gel onto a membrane, the method allows you to analyze and identify the DNA bands on a gel precisely. Your students will use Southern blotting to find a point mutation in the hemoglobin gene indicating Sickle Cell Anemia.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Southern blotting
Includes Instructions, Ready-to-Load™ DNA samples, agarose, electrophoresis buffer, nylon membranes, filter paper, blot stain
Requires Electrophoresis apparatus, power supply, microwave or hot plate, water bath and 80°C incubation oven
Time Range Electrophoresis: 45 min.; Blot: overnight; Stain and Destain: 10 min.
Content And Storage Some components require freezer storage
Class Size 5 Lab Groups
Product Type Electrophoresis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.