Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ InstaStain™ Blue
SDPSCI_ED

Catalog No. S05700 Shop All EDVOTEK Products
Change view
Click to view available options
Sufficient For:
100 Gels
40 Gels
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S05700 40 Gels
S05701 100 Gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05700 Supplier Edvotek™ Supplier No. 2003
Add to Cart
Add to Cart

Environmentally friendly, avoiding large amounts of liquid stain and waste disposal.

  • Sheets stain gels in minutes
  • High-quality, uniform gel stain with excellent results for photography

Specifications

Description Size: 7 x 7cm
Color Blue
Format Strips
Label or Dye Proprietary Mix
Number of Markers 0
Separation Range 14 to 200 kDa
Size Range 0.1 to 1 kb
Wells 8-well
Form Sheet, Solid
For Use With (Application) Excellent for photography
Quantity 7 x 7 cm, for 40 Gels
Sufficient For 40 Gels
Length (Metric) 7 cm
Width (Metric) 7 cm
Product Type DNA Stain
For Use With (Equipment) Mini Gel Tank
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.