Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ InstaStain™ Ethidium Bromide
SDPSCI_ED

Catalog No. S05698 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
7 x 7 cm, for 40 Gels
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05698 7 x 7 cm, for 40 Gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05698 Supplier Edvotek™ Supplier No. 2001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapid and sensitive.

  • Contains only a few micrograms of ethidium bromide
  • Gel is ready for visualization in two minutes
  • Much less ethidium bromide waste for disposal compared to standard staining procedure

Specifications

Color Dark Red/Purple
Format Strips
Label or Dye Ethidium Bromide
Form Sheet, Solid
For Use With (Application) Rapid and sensitive, disposal of InstaStain™ Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard Ethidium Bromide staining procedure
Quantity 7 x 7 cm, for 40 Gels
Sufficient For 40 Gels
Product Type Fluorescent DNA/RNA Stain
Stain Type Ethidium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.