Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Introduction to ELISA Reactions Kit
SDPSCI_ED

Catalog No. s90245nd Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90245ND 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S90245ND Supplier Edvotek™ Supplier No. 269
Only null left
Add to Cart
Edge
Add to Cart

Includes

  • Antigens
  • Primary antibody, Anti-IgG-peroxidase conjugate (secondary antibody)
  • Stabilized hydrogen peroxide
  • Peroxide co-substrate, ABTS substrate
  • Phosphate buffered saline concentrate
  • Two microtiter plates
  • Transfer pipets Microtest tubes with attached caps, 15mL plastic tubes

Requires

  • Distilled or deionized water
  • Beakers
  • 37°C incubation oven
  • Disposable lab gloves
  • Safety goggles
  • Automatic micropipets (0 to 50μL) and tips recommended

Teaches students the concepts of the Enzyme Linked Immunosorbent Assay (ELISA).

  • Determine the reaction steps of ELISA — a precise and sensitive antibody-based detection method for antigens such as bacterial and viral proteins
  • Perishable items shipped separately

No components have been prepared from human sources.

Order Info

Class size: 10 lab groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Teaches students the concepts of enzyme linked immunosorbent (elisa) assays
Includes Instructions, antigens, primary, secondary antibodies, peroxide co substrate, hydrogen peroxide, abts substrate, phosphate buffered saline, tubes, plates, and transfer pipets
Time Range 45 min.
Content And Storage Refrigerated; Store at: 4°C; 39°F
Class Size 10 Lab Groups
Product Type ELISA Reactions Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.