Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Ion-Exchange Chromatography Kit
SDPSCI_ED

Catalog No. s94617 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94617 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94617 Supplier Edvotek™ Supplier No. 243
Only null left
Add to Cart
Add to Cart

Includes

Materials for six separations: ion exchanger, CM-Cellulose; concentrated Potassium Acetate buffer, pH 6.0; dye mixture: blue dye, Cytochrome c; Cytochrome c Standard (10mg/mL); chromatography columns; plastic pipets; 1mL pipet; 15mL conical tubes; microcentrifuge tubes; complete instructions; background information; and study questions

Requires

5mL test tubes; ring stand with clamps for columns; 100 or 200mL beakers or flasks; 100, 250 and 500mL graduated cylinders; distilled water; 5mL pipets and pumps; and spectrophotometer and cuvets (optional, must be able to read 1mL samples)

Learn the principles of ion-exchange chromatography.

Most biological molecules have a net charge within a pH range of 2 to 10. When the pH is altered, the net charge on molecules can change drastically.

  • Learn the principles of ion-exchange chromatography by separating two charged molecules using a two-step salt gradient
  • Proteins are absorbed onto a charged solid support ion-exchange column and subsequently separated during elution under conditions that influence their net charge

Specifications

Grade 9 to Undergraduate
For Use With (Application) Learn The Principles of Ion-Exchange Chromatography
Includes Instructions, ion exchanger, chemical mixture, potassium acetate buffer, chromatography columns
Class Size 10 Lab Groups
Product Type Ion Exchange Chromatography Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.