Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Linking Food Science to Biotechnology: Unlock the Color of Candies Kit
SDPSCI_ED

Catalog No. S21247 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S21247 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S21247 Supplier Edvotek™ Supplier No. S47
Only null left
Add to Cart
Add to Cart

Investigate how agarose gel electrophoresis unlocks color code used by food scientists to make colorful candies.

  • Students extract color activities from common candies and separate dyes on agarose gel electrophoresis
  • Fun lab extension involves use of candy to build a DNA model
  • For 10 lab groups
  • Complete in 45 minutes
  • Includes: Instructions, dye extraction buffer, standard dye marker, UltraSpec-Agarose™ powder, 50X electrophoresis buffer, practice gel loading solution, transfer pipettes, plastic cups, microcentrifuge tube
  • Materials needed: Horizontal gel electrophoresis apparatus, power supply, automatic micropipettes with tips, microwave or hot plate, pipette pump, flasks or beakers
  • Storage: Stable at room temperature

Specifications

Grade 6 to 12
For Use With (Application) Fun lab extension involving use of candy to build a DNA model
Class Size 10 Lab Groups
Product Type Linking Food Science to Biotechnology: Unlock the Color of Candies Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.