Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Mini-Prep Isolation of Plasmid DNA
SDPSCI_ED

Catalog No. S05638 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S05638 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05638 Supplier Edvotek™ Supplier No. 202
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Plasmid LyphoCells™
  • Tris Buffer Concentrate
  • Sodium Hydroxide Solution
  • SDS Solution
  • Resuspension Buffer
  • Potassium Acetate Solution
  • RNase Solution
  • UltraSpec-Agarose™
  • 10X Gel Loading Solution
  • 50X Concentrated Electrophoresis Buffer
  • Practice Gel Loading Solution
  • FlashBlue™ DNA Stain
  • InstaStain™ Blue Cards
  • Disposable Pipets
  • Microcentrifuge Tubes.

Requires

  • Electrophoresis tank
  • Power supply
  • Variable micropipet and tips
  • Balance
  • Microwave or hotplate

Introduces the principles of extracting plasmid DNA from bacterial cells.

Small-scale rapid isolation of plasmid DNA is a routine procedure used for screening and analysis of recombinant DNA in cloning and subcloning experiments.

  • In this experiment, students isolate plasmid DNA without the use of toxic chemicals like phenol or chloroform
  • Complete in 1 hour 45 minutes

Order Info

For 20 plasmid isolations and seven 7x7cm gels.

Specifications

Format Kit
Isolation Technology Organic Extraction
For Use With (Equipment) Agarose Gel electrophoresis
For Use With (Application) Use for screening and analysis of recombinant DNA in cloning and subcloning experiments
Includes Instructions, plasmid extraction LyphoCells™, Gel-loading solution, UltraSpec-Agarose™ Powder, electrophoresis Buffer, InstaStain™ FlashBlue™ Stain, FlashBlue Plus™
Product Type Isolation of Plasmid DNA Kit
Quantity 1/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.