Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ PCR Beads
SDPSCI_ED

Catalog No. S96380 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
25 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96380 25 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96380 Supplier Edvotek™ Supplier No. 625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Beads for 25 reactions

Provide the reagents for Polymerase Chain Reactions (PCR) in a convenient, ambient-temperature-stable bead.

  • Beads for 25 reactions optimized for PCR; each bead contains Taq DNA polymerase, Taq DNA polymerase buffer, dNTP mixture and MgCl2
  • The only reagents that must be added to the reaction are template DNA and specific primers

Specifications

Grade 9 to Undergraduate
For Use With (Application) Provide the reagents for polymerase chain reactions (pcr) in a convenient ambient temperature stable bead
Includes Taq dna polymerase, taq dna polymerase buffer, dntp mixture, mgcl2
Quantity 25 Reactions
Class Size 25 Students
Product Type PCR Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.