Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Principles of Gel Filtration Chromatography
SDPSCI_ED

Catalog No. s11108 Shop All EDVOTEK Products
Change view
Click to view available options
Length (Metric):
5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S11108 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11108 Supplier Edvotek™ Supplier No. 108
Only null left
Add to Cart
Add to Cart

Includes

Instructions, sample mixture, chromatography columns, dry matrix, elution buffer, transfer pipets, microtest tubes

Teach the principles of gel filtration chromatography to your class.

  • Demonstrates how dyes of different colors separate on the basis of their size and shape
  • Columns may be rinsed and reused
  • Requires 50 or 100mL beakers, 25mL beaker or test tube and ring stands with clamps

Specifications

Diameter (Metric) Inner 1 cm
Type Chromatography Kit
Length (Metric) 5 in.
Includes Instructions, sample mixture, chromatography columns, dry matrix, elution Buffer, transfer pipets, microtest tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.