Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Protein InstaStain™ sheets
SDPSCI_ED

Catalog No. S94633 Shop All EDVOTEK Products
Change view
Click to view available options
Sufficient For:
15 Gels
30 Gels
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
S94633 15 Gels
S94634 30 Gels
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S94633 Supplier Edvotek™ Supplier No. 2016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stain gels faster than conventional methods avoiding large amounts of liquid stain and waste disposal.

Protein InstaStain™ sheets stain gels faster than conventional methods. Protein InstaStain™ gives high quality and uniform gel staining with excellent results for photography. They are also environmentally friendly because they use a solid matrix, avoiding large amounts of liquid stain and waste disposal. Protein InstaStain can be used to stain the most common gels of various sizes (ranging from 8x8 cm to 13x13 cm)

Order Info

Dimension: 10 x 10cm

Specifications

Color Blue
Format Strips
Label or Dye Coomassie
Number of Markers 0
Separation Range 14 to 200 kDa
Size Range 0.1 to 1 kb
Wells 8-well
Form Sheet, Solid
For Use With (Application) To stain Protein bands on Gel after electrophoresis
Quantity 15/Pk.
Sufficient For 15 Gels
Product Type Protein Stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.