Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Quantitative ELISA Laboratory Activity
SDPSCI_ED

Catalog No. S05654 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05654 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05654 Supplier Edvotek™ Supplier No. 278
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Instructions
  • Antigens
  • Primary and secondary antibodies
  • Substrate solution
  • Phosphate buffered saline
  • Blocking agent
  • Stop solution
  • Tubes
  • Plates
  • Transfer pipets

Requires

  • Distilled or deionized water
  • 37°C incubation oven
  • Automatic micropipets with tips
  • Laboratory glassware

Antibodies are highly specific in their recognition of antigens.

  • ELISA experiment demonstrates the quantification of varying concentrations of viral antigens as detected by the intensity of the color reaction due to the accumulation of products
  • Meets the requirements in the BSCS Blue Biology curriculum
  • Complete in two hours

All You Need:

  • distilled or deionized water
  • 37º C incubation oven
  • automatic micropipets with tips
  • laboratory glassware

Order Info

For six lab groups.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Demonstrates the quantitation of varying concentrations of viral antigens as detected by the intensity of the color reaction due to the accumulation of products
Includes Instructions, antigens, primary, secondary antibodies, substrate solution, phosphate buffered saline, blocking agent, stop solution, tubes, plates and transfer pipets
Time Range 2 Hr.
Content And Storage Refrigerated
Class Size 6 Lab Groups
Product Type ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.