Learn More
Edvotek Quick Plants™ Seeds
SDPSCI_ED
Catalog No. S11952 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
300 Seeds
50 Pk.
Scientific Name:
Arabidopis thaliana
Brassica
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Scientific Name
S11952 300 Seeds Arabidopis thaliana
S05696 50 Pk. Brassica
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S11952 Supplier Edvotek Supplier No. 1252
This item is not returnable. View return policy
From seed to flower in 6-7 weeks.
  • Grow rapidly and flourish under fluorescent light
  • Rapid growth make these plants ideal for classroom use
Specifications
For Use With (Application) Rapid growth make these plants ideal for classroom use.
Quantity 300 Seeds
Scientific Name Arabidopis thaliana
Product Type Seed, Wild Type Quick Plant