Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents for Electrophoresis: FlashBlue Plus DNA Staining System
SDPSCI_ED

Catalog No. S11609 Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11609 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11609 Supplier Edvotek™ Supplier No. 609
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

10x Concentrate for 1.2 L

Provides optimized solution to shorten time for staining and destaining steps.

  • For 1.2L

Specifications

Color Blue
Format Bottle
Form Liquid
For Use With (Application) Optimized to shorten the time required for both staining and destaining
Quantity 120 mL
Sufficient For 1.2L
Product Type DNA Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.