Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Reagents for Electrophoresis: Melt and Pour UltraSpec-Agarose™
SDPSCI_ED

Catalog No. S11601B Shop All EDVOTEK Products
Change view
Click to view available options
Quantity:
400 mL
5 x 400 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S11601B 5 x 400 mL
S11601 400 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S11601B Supplier Edvotek™ Supplier No. 601B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Complements the majority of EDVO-Kits that involve DNA electrophoresis - Edvotek™ Reagents for Electrophoresis: Melt and Pour UltraSpec-Agarose™ is sufficient for the separation of DNA fragments in the size range of 500-30,000 base pairs.

  • Melt and Pour UltraSpec-Agarose™ consists of 0.8% electrophoretic grade UltraSpec-Agarose™ in tris-acetate-EDTA buffer

Specifications

Buffer TAE
Color Brown
Concentration 0.8%
Format Melt and Pour
Gel Percentage 0.8%
Gel Size Mini
Separation of DNA
Separation Range 400 to ≥10,000 bp
Separation Type Electrophoresis
For Use With (Application) Agarose or Polyacrylamide Gel electrophoresis
Material Agarose
Quantity 5 x 400 mL
Product Type Agarose Gel
For Use With (Equipment) Mini Gel Tank
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.