Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Survey of Protein Diversity
SDPSCI_ED

Catalog No. S05635 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05635 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05635 Supplier Edvotek™ Supplier No. 150
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions; Denatured LyphoProtein™ samples; Standard protein markers; Gel-loading solution; Buffer; Protein InstaStain™

Requires

Three polyacrylamide gels (12%); MV10 vertical gel electrophoresis apparatus; Power supply; Hotplate or burner; Visualization (white light); Automatic micropipet with fine tips; Microtest tube holder; Lab glassware; Methanol; Glacial acetic acid; Distilled or deionized water

Polyacrylamide-based.

  • Learn about the diversity of proteins by studying the electrophoretic profiles of various sources.
  • Your students will separate proteins from bacterial, plant, serum, and milk proteins alongside a standard protein marker.
Experiment Times:
  • Electrophoresis, one hour
  • Staining, 20 minutes
  • Destaining, two hours

Order Info

For six groups sharing three gels.

Recommended Storage

Some components require freezer storage

Specifications

Grade 9 to Undergraduate
For Use With (Application) Separate proteins from bacterial, plant, serum, and milk proteins alongside a standard protein marker
Time Range 60 minutes; Staining time: 20 minutes
Content And Storage Refrigerated. Some components require freezer storage.
Class Size 6 Lab Groups
Product Type Protein Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.