Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ SYBR™ Safe DNA Stain
SDPSCI_ED

Catalog No. s05678 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05678 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S05678 Supplier Edvotek™ Supplier No. 608
Only null left
Add to Cart
Add to Cart

Improved staining efficiency without altering the DNA.

Used for post-electrophoresis DNA staining.

  • Displays no acute oral toxicity and greatly reduced genotoxicity
  • High sensitivity
  • Duration of staining: 10-15 min.
  • To visualize DNA expose gel to short/mid-range UV or Blue-Light Transilluminator
  • Non-mutagenic
  • More sensitive than ethidium bromide
  • Reduces waste storage and disposal costs
  • Offers non-hazardous waste status and Clean Water Act compliance
  • 10,000X Concentrate for 750 ml, Volume is 80 μl
Simple dilution: 1:10,000 (Add 5μL to 50mL of molten agar)

Specifications

Grade 9 to Undergraduate
For Use With (Application) For post-electrophoresis dna staining
Time Range 10 to 15 min.
Product Type Sybr™ Safe Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.