Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ The Mystery of the Crooked Cell
SDPSCI_ED

Catalog No. S96377 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96377 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96377 Supplier Edvotek™ Supplier No. S53
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Ready-to-Load™ dye samples
  • Agarose powder
  • Practice gel-loading solution
  • Electrophoresis buffer
  • Microtipped transfer pipets
  • Instructions

Requires

  • Electrophoresis tank
  • Power supply

Demonstrates detection of the mutation that causes sickle cell anemia.

Students use electrophoresis to separate dyes that represent patient samples and controls. Developed in partnership with Boston University School of Medicine.

Specifications

Grade 6 to 12
For Use With (Application) Demonstrates detection of the mutation that causes sickle cell anemia
Includes Instructions, ready-to-load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, microtipped transfer pipets
Quantity 10 Gels
Time Range 45 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.