Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Transformation of E. coli with Green Fluorescent Protein (GFP)
SDPSCI_ED

Catalog No. S68654 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68654 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68654 Supplier Edvotek™ Supplier No. 223/AP08
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

instructions, BactoBeads™ E. coli GFP Host, supercoiled pFluoroGreen™ plasmid DNA, ampicillin, IPTG, CaCl2, Growth Additive, ReadyPour™ Luria Broth Agar (sterile), Luria Broth Medium for Recovery (sterile), petri plates (small), petri plates (large), plastic microtipped transfer pipets, wrapped 10 ml pipet (sterile), toothpicks (sterile), inoculating loops (sterile), microcentrifuge tubes

GFP gene was isolated and cloned from the jellyfish Aequorea victoria.

In this experiment, students will explore the biological process of bacterial transformation using E. coli and plasmid DNA. At the end of the activity, students will have experience observing and analyzing acquired traits (ampicillin resistance and fluorescence) as exhibited by transformed bacterial cells.

Order Info

Contains materials for 10 transformations and controls.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Transformation of e. coli with a plasmid containing the gene for the green fluorescent protein (gfp)
Includes Instructions, lyphocells, e. coli gfp host, supercoiled pfluorogreen™, ampicillin, iptg, cacl2, readypour™ luria broth agar (sterile), luria broth medium for recovery (sterile), petri plates (small), petri plates (large
Time Range 15 min.
Content And Storage Refrigerated; Store at: -20°C; -4°F
Class Size 10 Lab Groups
Product Type Biotechnology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.