Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Transformation of E. coli with pGAL™ (Blue Colony)
SDPSCI_ED

Catalog No. S11293 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S11293 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S11293 Supplier Edvotek™ Supplier No. 221
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

BactoBeads, plasmid DNA, buffers, reagents and antibiotic; ReadyPour™ Luria Broth agar; Luria Broth media for recovery; 40 Petri plates; 20 plastic microtipped transfer pipets; Three wrapped 10mL pipets and 25 wrapped 1mL pipets; 20 inoculating loops; 40 microtest tubes with attached lids; Complete instructions, background information and study questions

Requires

Automatic micropipet (5 to 50μL) or 25μL fixed-volume micropipet and tips; Two water baths; Incubation oven; Pipet pumps or bulbs; Ice; Bunsen burner, hotplate or microwave oven; Hot gloves

Presence of IPTG is not required.

In this experiment, your students will develop an understanding of bacterial transformation by plasmid DNA by introducing an opportunity to observe an acquired phenotypic trait of the transformed bacterial cells. The presence of blue bacterial colonies visually demonstrates the expression of a specific gene for the Lac+ phenotype.

Order Info

Contains materials for 10 transformations.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Develop an understanding of bacterial transformation by plasmid dna
Time Range 50 min.
Content And Storage Some components require refrigeration or freezing
Class Size 10 Lab Groups
Product Type Transformation Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.