Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Western Blot Analysis, Polyacrylamide-Based
SDPSCI_ED

Catalog No. S90257 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S90257 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S90257 Supplier Edvotek™ Supplier No. 317
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Instructions
  • Negative control
  • All samples and antibodies
  • Various reagents and buffers
  • Membrane and filter paper

Requires

  • Three polyacrylamide gels (12%)
  • Vertical gel electrophoresis apparatus
  • Power supply
  • Automatic micropipet with fine tips
  • Laboratory glassware
  • Metric rulers
  • Distilled or deionized water
  • Glacial acetic acid
  • Methanol

In Western Blot analysis, protein identification is based on antibody and antigen reactions.

  • Proteins are separated on a polyacrylamide gel and are transferred (blotted) to a nylon membrane
  • Membrane is exposed to solutions containing primary antibody, followed by a secondary antibody coupled to an enzyme
  • Membrane is then soaked in a substrate solution to develop the color reaction to identify the antigen protein band
  • Molecular weights of the visible bands are measured using prestained protein markers of known molecular weight

Experiment times:

  • Electrophoresis, one hour
  • Blot, overnight
  • Detection, two hours and 30 minutes

Kit does not require an electrotransfer apparatus.

Order Info

Contains materials for six blots.

Specifications

Grade 9 to Undergraduate
For Use With (Application) Proteins are separated on polyacrylamide gels and are transferred (blotted) to a nylon membrane
Includes Instructions, negative control, all samples and antibodies, various reagents and buffers, membrane and filter paper
Quantity 6 blots
Time Range 2.5 hr.
Content And Storage Refrigerated
Class Size 6 Lab Groups
Product Type Western Blot Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.