Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ What is an Epidemic and how does an Infection Spread?
SDPSCI_ED

Catalog No. s94602 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94602 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This kit contains chemical(s) that may be harmful if used improperly. This kit is to be used by students only with teacher supervision.
Catalog No. S94602 Supplier Edvotek™ Supplier No. S68
Only null left
Add to Cart
Add to Cart
This kit contains chemical(s) that may be harmful if used improperly. This kit is to be used by students only with teacher supervision.

Includes

  • Sodium hydroxide pellets
  • 1N HCl
  • Color indicator
  • Test tubes and caps
  • Large transfer pipets
  • Complete instructions, background information and study questions

Requires

  • Gloves
  • Safety goggles
  • Lab coats or protective clothing

Students develop skills for accurately transferring solutions using pipets.

This experiment contains solutions to demonstrate how an infection is spread in a population through contact with simulated body fluids.

  • Learn how infections are spread from one person to many others
  • Transfer solutions from one tube to another
  • Test a solution for the presence of an acid or a base

Order Info

Class size: 10 groups.

Specifications

Grade 6 to 12
For Use With (Application) Infections are spread from one person to many others, transfer solutions from one tube to another, test a solution for the presence of an acid or a base
Includes Instructions, hcl solution, naoh, color indicator, test tubes and pipets
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type What Is An Epidemic and How Does An Infection Spread Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.