Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ What Is PCR and How Does It Work?
SDPSCI_ED

Catalog No. S68633 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68633 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68633 Supplier Edvotek™ Supplier No. S48
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Dye samples
  • Practice gel-loading solution
  • UltraSpec-Agarose™ powder
  • Concentrated electrophoresis buffer
  • Transfer pipets
  • Instructions and study questions

Requires

  • Horizontal gel electrophoresis apparatus
  • DC power supply
  • Microwave, hotplate or burner

Demonstrates the process of DNA amplification by PCR.

  • Polymerase Chain Reaction (PCR) has played an important role in the advancement of molecular biology and the enhancement of DNA isolation, detection and amplification
  • Simulation demonstrates the process of DNA amplification by PCR and how the amplified product is detected by separating the reaction mixture by agarose gel electrophoresis

Order Info

Class size: 10 groups.

Specifications

Grade 6 to 12
For Use With (Application) Demonstrates the process of dna amplification by pcr and detected by separating the reaction mixture by agarose gel electrophoresis
Includes Instructions, ready-to-load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, microtipped transfer pipets
Time Range 45 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.