Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Whose DNA Was Left Behind? Experiment Kit
SDPSCI_ED

Catalog No. S94591 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94591 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94591 Supplier Edvotek™ Supplier No. S51
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Instructions, Ready-to-Load QuickStrip™ Dye Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, Disposable Pipets

Requires

Electrophoresis apparatus, DC power source, microwave, hotplate or burner, beakers (250mL), beaker tongs or hot gloves, distilled water, balance

Students simulate DNA fingerprinting and use evidence to narrow down a list of criminal suspects.

Introduces the basics of DNA fingerprinting and agarose gel electrophoresis.

  • Introductory-level exercise with hands-on experimentation
  • Illustrates how DNA sequences can be isolated in trace evidence such as blood cells or hair
  • Dye simulations function as DNA samples for analysis
  • Class size: 10 groups

Order Info

Results must be analyzed upon completion and cannot be stored overnight.

Specifications

Grade 6 to 12
For Use With (Application) Use dna fingerprinting to compare simulated crime scene dna with that of two suspects and try to catch the criminal
Includes Instructions, ready-to-load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, microtipped transfer pipets
Time Range 45 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.