Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Whose Fingerprints Were Left Behind? Experiment Kit
SDPSCI_ED

Catalog No. S94590 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S94590 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S94590 Supplier Edvotek™ Supplier No. S91
Add to Cart
Add to Cart

Includes

instructions, brushes, magnifying lens, fingerprint cards, black dusting powder, grey dye dusting powder, fingerprint lifters

Requires

Alcohol, UV longwave light source, safety goggles, scissors, towel/washcloth

Find fingerprints invisible to the naked eye.

Students solve a crime by dusting for fingerprints and use fluorescent dust to search for and identify trace amounts of blood.

  • Demonstrates the proper dusting and lifting techniques to collect latent fingerprints at a crime scene
  • Learn how to identify microscopic amounts of blood with a fluorescent light
  • Class size: 10 groups
  • Run time: 50 minutes

Specifications

Grade 6 to 12
For Use With (Application) Demonstrates dusting, lifting techniques to collect fingerprints at crime scene
Includes Instructions, brushes, magnifying lens, fingerprint cards, black dusting powder, fluorescent green and grey dye dusting powder, fingerprint lifters
Time Range 50 min.
Content And Storage Room temperature
Class Size 10 Lab Groups
Product Type Classroom Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.