Learn More
Ideal Vacuum Products LLC Edwards 12 RV12 Vacuum Pump Major Repair Rebuild Kit, Including Vanes
SDP
Catalog No. NC1242686
Encompass
These Edwards RV12 vacuum pump major rebuild kit includes coupling elements, o'rings, gaskets, airbleed assembly, ducting sheet, shaft seals, vanes, and other parts needed for major service. These Edwards RV12 vacuum pump repair rebuild kits are guaranteed to meet or exceed OEM specifications.
Catalog No. NC1242686 Supplier Ideal Vacuum Products LLC Supplier No. P101886
These Edwards RV12 vacuum pump major rebuild kit includes coupling elements, o'rings, gaskets, airbleed assembly, ducting sheet, shaft seals, vanes, and other parts needed for major service. These Edwards RV12 vacuum pump repair rebuild kits are guaranteed to meet or exceed OEM specifications.