Learn More
Precision Plus Vacuum Parts Inc Edwards, E2M28,E2M30 Maintenance kit
Catalog No. 501539876
Encompass_Preferred
Edwards, E2M28,E2M30, Maintenance kits includes vanes, springs and pins (as required) seals, coupling element, o-rings, and gaskets
Catalog No. 50-153-9876 Supplier Precision Plus Vacuum Parts Inc Supplier No. 37301800ED
Edwards, E2M28,E2M30, Maintenance kits includes vanes, springs and pins (as required) seals, coupling element, o-rings, and gaskets