Learn More
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange
Catalog No. 1128317 Shop All Edwards High Vacuum International Products
Change view
Click to view available options
Type:
NW25 Blank Flange
Catalog No. Type
11-283-17 NW25 Blank Flange
1 options
This item is not returnable. View return policy
Catalog No. 11-283-17 Supplier Edwards Supplier No. C10514368
This item is not returnable. View return policy
For use with BOC high vacuum pumps
  • Made of aluminum
  • Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
For Use With BOC high vacuum pump
Material Aluminum
Size 40 x 26.2 x 6 x 2.5 mm
Type NW25 Blank Flange