Learn More
Edwards Fittings for BOC High Vacuum Pumps: Centering Ring
Catalog No. 1128331 Shop All Edwards High Vacuum International Products
Change view
Click to view available options
Material:
Polymer
Stainless Steel
Type:
NW16 Viton Centering Ring
NW25/25 Viton Centering Ring
NW40 Centering Ring
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Type
11-283-31 Polymer NW25/25 Viton Centering Ring
11-282-30 Polymer NW16 Viton Centering Ring
11-283-32 Stainless Steel NW40 Centering Ring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-283-31 Supplier Edwards Supplier No. C10514395
Add to Cart
Add to Cart
For use with BOC high vacuum pumps
  • Aluminum centering rings manufactured from high-tech polymer
  • Unique slotted design prevents gas bursts
  • CX2 polymer can be used at temperatures up to 120°C and is unaffected by most common solvents
  • Outgassing rate of the material is 6.6 × 10-8mbar l s-1cm-2
Specifications
For Use With BOC high vacuum pump
Material Polymer
Size 3.9 x 8 mm
Type NW25/25 Viton Centering Ring