Learn More
Precision Plus Vacuum Parts Inc Edwards, RV8, Major Repair Kit
Catalog No. 501539885
Encompass_Preferred
Edwards, RV8, Major Repair Kit with blades, springs, pins and coupling elements, o-rings,gaskets, airbleed assembly, ducting sheet, shaft seals.
Catalog No. 50-153-9885 Supplier Precision Plus Vacuum Parts Inc Supplier No. 65201131RV8
Edwards, RV8, Major Repair Kit with blades, springs, pins and coupling elements, o-rings,gaskets, airbleed assembly, ducting sheet, shaft seals.