Learn More
PIE SCIENTIFIC LLC EDWARDS NXDS10I DRY PUMP
SDP
Catalog No. NC2186731
Encompass_Preferred
  • Oil-free vacuum environment prevents contamination and eliminates oil disposal concerns.
  • Low noise level operation of 52 dB(A) improves the working environment.
  • Efficient vapor handling with gas ballast expands application range.
  • Extended service intervals due to advanced tip seal technology.
  • Optimized performance and flexibility with intelligent control features.
  • Compact design for easy integration into various lab settings.
  • Operates within a temperature range of 10 to 40 °C (41 to 104 °F).
  • Lightweight at 25.8 kg (57 lb) for easy handling.
  • Integrated inverter drive for energy-efficient operation.
This item is not returnable. View return policy
Catalog No. NC2186731 Supplier PIE SCIENTIFIC LLC Supplier No. A202
Add to Cart
Add to Cart
Request a Quote
  • Oil-free vacuum environment prevents contamination and eliminates oil disposal concerns.
  • Low noise level operation of 52 dB(A) improves the working environment.
  • Efficient vapor handling with gas ballast expands application range.
  • Extended service intervals due to advanced tip seal technology.
  • Optimized performance and flexibility with intelligent control features.
  • Compact design for easy integration into various lab settings.
  • Operates within a temperature range of 10 to 40 °C (41 to 104 °F).
  • Lightweight at 25.8 kg (57 lb) for easy handling.
  • Integrated inverter drive for energy-efficient operation.