Learn More
Edwards Rotary Vane Vacuum Pump Fittings: Clamping Ring
Catalog No. 1128389 Shop All Edwards High Vacuum International Products
Change view
Click to view available options
For Use With:
Rotary vane vacuum pump
Catalog No. For Use With
11-283-89 Rotary vane vacuum pump
1 options
Catalog No. 11-283-89 Supplier Edwards Supplier No. C10514401
Range of coupling clamps manufactured from high technology polymer for vacuum system builder
  • Clamping ring: compact stainless-steel
  • Polymer clamp: lightweight, neat appearance
  • Aluminum clamp: rugged Pneurop standard
  • Metal clamp: suitable for aluminum and indium seals — wide temperature range
  • Claw clamps and bolts: wide range optimized for many applications — high strength CF bolts for UHV flanges
Specifications
For Use With Rotary vane vacuum pump
Material Stainless Steel
Type Clamping Ring