Learn More
Electron Microscopy Sciences Edwards RV5 Vacuum Pump, 220 V
SDP
Catalog No. 5025710
Encompass
Rotary vacuum pump with double stage. 4.1 cfm (117L/min) displacement. Ultimate vacuum (without gas ballast) 1.5x10-3 torr. Max. inlet pressure for water vapor: 38 torr. Max. water vapor pumping rate: 0.48 lb/h. Motor: 1⁄2 hp. Oil capacity Max/Min 0.7L/0.42L.
Catalog No. 50-257-10 Supplier Electron Microscopy Sciences Supplier No. 91005E
Rotary vacuum pump with double stage. 4.1 cfm (117L/min) displacement. Ultimate vacuum (without gas ballast) 1.5x10-3 torr. Max. inlet pressure for water vapor: 38 torr. Max. water vapor pumping rate: 0.48 lb/h. Motor: 1⁄2 hp. Oil capacity Max/Min 0.7L/0.42L.