Learn More
Electron Microscopy Sciences Edwards RV8 Vacuum Pump, 110 V
SDP
Catalog No. 5025712
Encompass
Rotary vacuum pump with double stage. 6.9 cfm (195 L/min) displacement. Ultimate vacuum (without gas ballast) 1.5x10-3 torr. Max. inlet water pressure vapor: 29 torr. Max. water vapor pumping rate: .48 lb/h. Motor: 3⁄4 hp. Oil capacity Max/Min 0.75L/0.43.
Catalog No. 50-257-12 Supplier Electron Microscopy Sciences Supplier No. 91025
Add to Cart
Add to Cart
Request a Quote
Rotary vacuum pump with double stage. 6.9 cfm (195 L/min) displacement. Ultimate vacuum (without gas ballast) 1.5x10-3 torr. Max. inlet water pressure vapor: 29 torr. Max. water vapor pumping rate: .48 lb/h. Motor: 3⁄4 hp. Oil capacity Max/Min 0.75L/0.43.