Learn More
Across International Edwards RV8 6.9 CFM Dual-Stage Vacuum Pump with Bellow & Fitting
SDP
Catalog No. NC1589581
Encompass_Preferred
Edwards RV8 6.9 CFM Dual-Stage Vacuum Pump with Bellow & Fitting
Catalog No. NC1589581 Supplier Across International Supplier No. RV8
Add to Cart
Add to Cart
Request a Quote
Edwards RV8 6.9 CFM Dual-Stage Vacuum Pump with Bellow & Fitting