Learn More
Electron Microscopy Sciences Edwards Ultragrade 19 1 LT
SDP
Supplier:  Electron Microscopy Sciences 60740
Encompass_Preferred
Edwards Ultragrade 19 Oil is specially recommended for use in clean pumping applications. The oil offers reduced maintenance cost, longer fluid life, and a cooler running pump.
Catalog No. 50-996-276