Learn More
Electron Microscopy Sciences Edwards Ultragrade 19 4 LT
SDP
Catalog No. 50996277
Encompass_Preferred
Edwards Ultragrade 19 Oil is specially recommended for use in clean pumping applications. The oil offers reduced maintenance cost, longer fluid life, and a cooler running pump.
Catalog No. 50-996-277 Supplier Electron Microscopy Sciences Supplier No. 60742
Edwards Ultragrade 19 Oil is specially recommended for use in clean pumping applications. The oil offers reduced maintenance cost, longer fluid life, and a cooler running pump.