Learn More
Scientific Instrument Services Inc Edwards ULTRAGRADE™ 19 Oil 12 X 1L
Catalog No. NC9734245
Encompass
Edwards ULTRAGRADE™ Performance 19 uses a solvent refining process, whereas this oil is produced by a unique hydro treating process. This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities. When combined with additives, the ULTRAGRADE™ Performance oils give improved vacuum performance, thermal and oxidation stability and rust inhibition. The properties of the oils allow them to be used in a wide range of applications. 12x1L
Catalog No. NC9734245 Supplier Scientific Instrument Services Inc Supplier No. H11025012
Edwards ULTRAGRADE™ Performance 19 uses a solvent refining process, whereas this oil is produced by a unique hydro treating process. This process eliminates nitrogen, sulphur and oxygen and the aromatic hydrocarbons, leaving a clear base stock essentially free of impurities. When combined with additives, the ULTRAGRADE™ Performance oils give improved vacuum performance, thermal and oxidation stability and rust inhibition. The properties of the oils allow them to be used in a wide range of applications. 12x1L