Learn More
Electron Microscopy Sciences MAC Microanalysis Microanalysis Standard NBS SRM 481
SDP
Supplier:  Electron Microscopy Sciences 8007601
Encompass_Preferred
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. All of our standards are polished to a 0.25 micron diamond finish and carbon coated. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Specify 25 mm or 32 mm holder. Includes NBS glass SRM 481 Au/Ag set of 6 standards, comprises 4 alloys and 2 pure metals.
Catalog No. 50-274-61