Learn More
Electron Microscopy Sciences MAC Microanalysis Standard SRM 710 Grain, 2 x 2 mm
SDP
Supplier:  Electron Microscopy Sciences 80075NBS710
Encompass_Preferred
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Grain of standard measures approximately 2mm x 2mm in size.
Type: Grain
Size: 2 x 2 mm
Catalog No. 50-263-46