Learn More
Electron Microscopy Sciences MAC Microanalysis Standard Titanium Mount, 2, 3, or 5 mm
SDP
Supplier:  Electron Microscopy Sciences 80074TI
Encompass_Preferred
All of our standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems. They are supplied with fully authenticated certificates of analyses and a location map for standard identification. Mounts are polished to 0.25 micron diamond and carbon coated. Supplied in 2, 3, or5 mm brass tube.
Type: Mounted
Size: 2, 3, or 5 mm
Catalog No. 50-266-66