Learn More
General Laboratory Products EE BROTH MOSSEL 100 ML 12/CS
Catalog No. 50855957
Encompass_Preferred
Media; Microbiology; Bottle; EE Broth Mossel; Selective enrichment for Enterobacteriaceae, particularly from foods; Wheaton Easy Pour Bottle; Size: 100mL; 12/case
Catalog No. 50-855-957 Supplier General Laboratory Products Supplier No. 8WM5000100
Media; Microbiology; Bottle; EE Broth Mossel; Selective enrichment for Enterobacteriaceae, particularly from foods; Wheaton Easy Pour Bottle; Size: 100mL; 12/case