Learn More
NEOGEN Enterobacteria Enrichment (EE) Broth Mossel, 10 kg
SDP
Catalog No. 502015320
Encompass_Preferred
Enterobacteria Enrichment (EE) Broth Mossel is used for the cultivation and enrichment of Enterobacteriaceae in food. Enterobacteria Enrichment (EE) Broth Mossel, conforms to Harmonized USP/EP/JP Requirements and is not intended for use in the diagnosis of disease or other conditions in humans. Enterobacteria Enrichment Broth Mossel was developed by Mossel, Visser, and Cornelissen to facilitate the growth of Enterobacteriaceae. This medium contains dextrose to enhance the growth of E. coli and Salmonella spp., particularly in food samples. Nuisance organisms are suppressed by the addition of Ox Bile and Brilliant Green. Enterobacteria Enrichment Broth Mossel is used as an enrichment broth, providing a rich environment for the recovery of damaged or injured cells.For more information visit https://www.neogen.com/solutions/Microbiology/enterobacteria-enrichment-broth-mossel
Catalog No. 50-201-5320 Supplier NEOGEN Supplier No. 700003127
Enterobacteria Enrichment (EE) Broth Mossel is used for the cultivation and enrichment of Enterobacteriaceae in food. Enterobacteria Enrichment (EE) Broth Mossel, conforms to Harmonized USP/EP/JP Requirements and is not intended for use in the diagnosis of disease or other conditions in humans. Enterobacteria Enrichment Broth Mossel was developed by Mossel, Visser, and Cornelissen to facilitate the growth of Enterobacteriaceae. This medium contains dextrose to enhance the growth of E. coli and Salmonella spp., particularly in food samples. Nuisance organisms are suppressed by the addition of Ox Bile and Brilliant Green. Enterobacteria Enrichment Broth Mossel is used as an enrichment broth, providing a rich environment for the recovery of damaged or injured cells.For more information visit https://www.neogen.com/solutions/Microbiology/enterobacteria-enrichment-broth-mossel