Learn More
General Laboratory Products EE BROTH MOSSEL 90 ML 12/CS
Catalog No. 50855956
Encompass_Preferred
Media; Microbiology; Bottle; EE Broth Mossel; Selective enrichment for Enterobacteriaceae, particularly from foods; Wheaton Easy Pour Bottle; Wide mouth w/screw cap; Size: 90mL; 12/case
Catalog No. 50-855-956 Supplier General Laboratory Products Supplier No. 8WM500090
Media; Microbiology; Bottle; EE Broth Mossel; Selective enrichment for Enterobacteriaceae, particularly from foods; Wheaton Easy Pour Bottle; Wide mouth w/screw cap; Size: 90mL; 12/case