Learn More
General Laboratory Products EE Broth Mossel, Selective Enrichment Medium For Enterobacteriaceae, 900 mL
Catalog No. NC2151047
Encompass_Preferred
EE Broth Mossel is a selective enrichment medium that supports the growth of Enterobacteriaceae, including *E. coli* and *Salmonella*, from food samples. Ideal for food microbiology, it utilizes selective agents like ox bile and brilliant green to inhibit unwanted flora, ensuring reliable results in bacteriological examinations. This broth ensures quality compliance with Harmonized USP/EP/JP standards, distinguishing it in its category as a robust option for critical microbiological testing.
This item is not returnable. View return policy
Catalog No. NC2151047 Supplier General Laboratory Products Supplier No. 1000RPC5000900
EE Broth Mossel is a selective enrichment medium that supports the growth of Enterobacteriaceae, including *E. coli* and *Salmonella*, from food samples. Ideal for food microbiology, it utilizes selective agents like ox bile and brilliant green to inhibit unwanted flora, ensuring reliable results in bacteriological examinations. This broth ensures quality compliance with Harmonized USP/EP/JP standards, distinguishing it in its category as a robust option for critical microbiological testing.