Learn More
MOLTOX Molecular Toxicology Inc EE Mossel Broth (Enterobacteriaceae Enrichment Broth), 90 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948759
Encompass_Preferred
EE Mossel Broth is used for selectively enriching and detecting Enterobacteriaceae and other lactose-negative organisms. This is then followed by subculturing on a selective medium such as Violet Red Bile or Viloet Red Bile Glucose Agar (VRBGA). Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-759 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26554.038
EE Mossel Broth is used for selectively enriching and detecting Enterobacteriaceae and other lactose-negative organisms. This is then followed by subculturing on a selective medium such as Violet Red Bile or Viloet Red Bile Glucose Agar (VRBGA). Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.